Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)
TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer), Glass bottle, 1 L
Catalog # NADIEC-860-1L
Marketsource
Supplier: NATIONAL DIAGNOSTICS
Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)
Catalog # NADIEC-860-1L
Supplier: NATIONAL DIAGNOSTICS
Marketsource
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Description:TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer)
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



