Tween® 20 (Polysorbate)
Glass bottle, 200 mL
Catalog # NADIEC-607-200ML
Marketsource
Supplier: NATIONAL DIAGNOSTICS
Tween® 20 (Polysorbate)
Catalog # NADIEC-607-200ML
Supplier: NATIONAL DIAGNOSTICS
Marketsource
CAS Number: 9005-64-5
Jump to:
Product Details & Documents
Synonyms: Polyoxyethylene sorbitan monolaurate
- CAS Number:9005-64-5
- Formula:C₅₈H₁₁₄O₂₆
- MDL Number:MFCD00165986
- Molecular Weight:1227.53 g/mol
- Density:1.1 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:500-018-3
Specifications
Specifications
- Pack type:Glass bottle
- Pk:200 mL
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



