Pycnometer heads with/without bottle
1000 ml, 60/37
Catalog # LENZ05660007
Supplier: Lenz Laborglas
Jump to:
Product Details & Documents
Set, made of DURAN® borosilicate glass, consists of wide neck bottle with conical shoulder and NS joint, and pycnometer head.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Suitable for density determinations of bitumen-like and mineral materials.
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
Specifications
- NS:60/37
- Volume (nom.):1000 ml
- Seq:02
Accessories
Product Information
- Catalog #AvailabilityPrice
- Bottle for pycnometer NS60/46, 1000 ml