Orders LinkContactUsLinkComponent
Separating funnels, Gilson
Separating funnels, Gilson
100 ml, 19/26 NS
Catalog # 527-1507
Supplier:   Lenz Laborglas
Separating funnels, Gilson
Catalog # 527-1507
Supplier:  Lenz Laborglas
Separating funnels, Gilson
Separating funnels, Gilson
100 ml, 19/26 NS
Price
SFr. 154.00
Log in for availability
1 * 1 ST
Product Details & Documents
DURAN®, borosilicate glass, with NS-PE stopper.
With socket and plain stem. Available with interchangeable solid glass key or alternatively PTFE key for grease-free performance. Without graduation.
Documents
Specifications
  • Socket size:
    19/26 NS
  • Capacity:
    100 ml