Orders LinkContactUsLinkComponent
Separating funnels, Gilson
Separating funnels, Gilson
500 ml, 29/32 NS
Catalog # 527-1504
Supplier:   Lenz Laborglas
Separating funnels, Gilson
Catalog # 527-1504
Supplier:  Lenz Laborglas
Separating funnels, Gilson
Separating funnels, Gilson
500 ml, 29/32 NS
Price
SFr. 163.00
Log in for availability
1 * 1 ST
Product Details & Documents
DURAN®, borosilicate glass, with NS-PE stopper.
With socket and plain stem. Available with interchangeable solid glass key or alternatively PTFE key for grease-free performance. Without graduation.
Documents
Specifications
  • Socket size:
    29/32 NS
  • Capacity:
    500 ml