Separating funnels, Gilson
250 ml, 29/32 NS
527-1503
DURAN®, borosilicate glass, with NS-PE stopper.
With socket and plain stem. Available with interchangeable solid glass key or alternatively PTFE key for grease-free performance. Without graduation.
- Socket size:29/32 NS
- Capacity:250 ml