Orders LinkContactUsLinkComponent
Separating funnels, Squibb
Separating funnels, Squibb
50 ml, 19/26 NS, -
Catalog # 527-1331
Supplier:   Lenz Laborglas
Separating funnels, Squibb
Catalog # 527-1331
Supplier:  Lenz Laborglas
Separating funnels, Squibb
Separating funnels, Squibb
50 ml, 19/26 NS, -
Price
SFr. 88.80
Log in for availability
1 * 1 ST
Product Details & Documents
DURAN®, borosilicate glass, with NS-PE stopper.
With socket and plain stem. Available with interchangeable solid glass key or alternatively PTFE key for grease-free performance. With or without graduations.
Documents
Specifications
  • Socket size:
    19/26 NS
  • Capacity:
    50 ml
  • Division:
    -