Stopcocks, one-way
10** mm, 29,2 NS
Catalog # 227-3140
Supplier: Lenz Laborglas
Jump to:
Product Details & Documents
DURAN®, borosilicate glass, with solid glass or PTFE key.
- Replaceable NS keys
- Screw thread retaining device
- Leakage rate compliant with DIN 12540-1
Straight bore.
* with capillary side arms
** with rubber retaining device.
Certifications: DIN 12554.
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
Specifications
- Hole Ø:10** mm
- Seq:10
- Spigot:29,2 NS