Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Slide trays

Slide trays

Slide tray, White, PVC, 680×190 mm, 40
Catalog # 631-9144
Supplier:   KARTELL
Slide trays
Catalog # 631-9144
Supplier:  KARTELL
Slide trays
Slide trays

Slide tray, White, PVC, 680×190 mm, 40

Price
SFr. 39.90
Log in for availability
1 * 1 ST
Product Details & Documents

Identification of different dyes on the slides.

  • Easy removal of slides
  • Slide trays for 76×26 mm slides

Colour: White

Specifications
  • No. of slides:
    40
  • W×D:
    680×190 mm
  • Colour:
    White
  • Description:
    Slide tray
  • Material:
    PVC
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.