Orders LinkContactUsLinkComponent
Weighing bottles, PP
Weighing bottles, PP
60 ml, 40×69 mm
Catalog # 611-0104
Supplier:   KARTELL
Weighing bottles, PP
Catalog # 611-0104
Supplier:  KARTELL
Weighing bottles, PP
Weighing bottles, PP
60 ml, 40×69 mm
Price
SFr. 7.40
Log in for availability
1 * 1 ST
Product Details & Documents
PP with lid.
  • Very good chemical resistance
  • Very good temperature resistance
Cylindrical shape, with knobbed lid.
Specifications
  • Ø×H:
    40×69 mm
  • Capacity:
    60 ml