Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Sample Cups for Analysers

For Centrichem® analysers, 0,25 ml, 16,4 mm, 13,7 mm
  391-1017
 :   KARTELL
Sample Cups for Analysers
  391-1017
 :  KARTELL
Sample Cups for Analysers
Sample Cups for Analysers

For Centrichem® analysers, 0,25 ml, 16,4 mm, 13,7 mm

 
SFr. 44.90
   
1 * 1.000 ST
 

PS, transparent.

 : *The product(s) on this page are outside the scope of the EU Directive 2019/904

 
  • Ext.Ø:
    13,7 mm
  • Capacity:
    0,25 ml
  • Description:
    For Centrichem® analysers
  • Height:
    16,4 mm
  • Seq:
    01