Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Erlenmeyer flasks, narrow neck, with standard socket

Erlenmeyer flasks, narrow neck, with standard socket

500 ml, 50 ml, 24/29 NS
Catalog # 214-0135
Supplier:   KARTELL
Erlenmeyer flasks, narrow neck, with standard socket
Catalog # 214-0135
Supplier:  KARTELL
Erlenmeyer flasks, narrow neck, with standard socket
Erlenmeyer flasks, narrow neck, with standard socket

500 ml, 50 ml, 24/29 NS

Price
SFr. 12.70
Log in for availability
1 * 1 ST
Product Details & Documents

PP, transparent.

  • Very good chemical resistance
  • Resistant to heat up to 121 °C (for 20 minutes), prolonged use 100 °C
  • Smooth surfaces for easy, fast cleaning

With moulded graduations. Autoclavable. Supplied without stoppers.

Specifications
  • Graduation Interval:
    50 ml
  • Socket size:
    24/29 NS
  • Capacity:
    500 ml