Order Entry
Switzerland
Orders LinkContactUsLinkComponent
n-Hexane, anhydrous

n-Hexane, anhydrous

Glass bottle, 1 L
Catalog # 43263.K2
Flammable
Health
Irritant
Environment
n-Hexane, anhydrous
Catalog # 43263.K2
CAS Number:  110-54-3
n-Hexane, anhydrous
n-Hexane, anhydrous

Glass bottle, 1 L

CAS Number:  110-54-3
Price
289.00 SFr.
Log in for availability
1 * 1 L
Product Details & Documents

CAS No.: 110-54-3 Molecular Formula: C6H14 Formula Weight: 86.18 Flash Point: -23°(-9°F) Physical Form: Liquid Density: 0.659 Refractive Index: 1.3750 Appearance: Colourless Odour: Weak, characteristic Melting Point: -95° Boiling Point: 69° Beilstein Reference: 1730733 Merck Reference: 14,4694 UN No.: 1208 MDL No.: MFCD00009520

Solvent in Sol-Gel precursor applications

Synonyms:  Hexyl hydride, Hexanen, Hexane
  • CAS Number:110-54-3
  • Formula:H₃C(CH₂)₄CH₃
  • MDL Number:MFCD00009520
  • Molecular Weight:86.18 g/mol
  • Merck Index:14,4694
  • Boiling Pt:69 °C
  • Melting Pt:–95 °C
  • Density:0.659
  • Flash Pt:–23 °C (–9 °F)
  • Storage Temperature:Ambient
  • EINECS:203-777-6
  • UN:1208
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.