Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Propionic acid 99+%, extra pure

Propionic acid 99+%, extra pure

Glass bottle, 1 L
Catalog # ACRO220130010
Corrosive
Flammable
Irritant
Propionic acid 99+%, extra pure
Catalog # ACRO220130010
CAS Number:  79-09-4
Propionic acid 99+%, extra pure
Propionic acid 99+%, extra pure

Glass bottle, 1 L

CAS Number:  79-09-4
Price
SFr. 97.10
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Propanoic acid
  • CAS Number:79-09-4
  • Formula:C₃H₆O₂
  • MDL Number:MFCD00002756
  • Molecular Weight:74.08 g/mol
  • Merck Index:13,07917
  • Boiling Pt:141 °C (1013 hPa)
  • Melting Pt:–22 °C
  • Density:0.99 g/cm³
  • Flash Pt:51 °C
  • Storage Temperature:Ambient
  • EINECS:201-176-3
  • UN:3463
  • ADR:8,II
  • REACH:01-2119486971-24

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3850 to 1.3870 (20°C, 589 nm)
  • Water:
    ≤0.1 %
  • GC:
    ≥99.0 %
  • Color scale:
    ≤10 APHA

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.