Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Vanadium(V) oxide 98+%

Vanadium(V) oxide 98+%

Plastic bottle, 500 g
Catalog # ACRO207995000
Corrosive
Irritant
Health
Environment
Vanadium(V) oxide 98+%
Catalog # ACRO207995000
CAS Number:  1314-62-1
Vanadium(V) oxide 98+%
Vanadium(V) oxide 98+%

Plastic bottle, 500 g

CAS Number:  1314-62-1
Price
SFr. 164.00
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Vanadium pentoxide, di-Vanadium pentaoxide
  • CAS Number:1314-62-1
  • Formula:O₅V₂
  • MDL Number:MFCD00011457
  • Molecular Weight:181.88 g/mol
  • Merck Index:13,09987
  • Boiling Pt:1750 °C (760 mmHg)
  • Melting Pt:690 °C
  • Density:3.35 g/cm³
  • Storage Temperature:Ambient
  • EINECS:215-239-8
  • UN:2862
  • ADR:6.1,III

Documents

Specifications

Specification Test Results

  • Appearance:
    Yellow to orange-brown Powder
  • Assay:
    ≥98.0 %
  • Particle size:
    ≤500 µm
  • Chromium (Cr):
    ≤200 ppm
  • Iron (Fe):
    ≤1000 ppm
  • Silicon (Si):
    ≤200 ppm
  • Sodium oxide (Na2O):
    ≤300 ppm
  • Potassium oxide (K2O):
    ≤300 ppm

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 g

Specifications