Nonadecanoic acid 98+%
Glass bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
Nonadecanoic acid 98+%
Catalog # ACRO207190050
Supplier: Thermo Fisher Scientific
CAS Number: 646-30-0
Jump to:
Product Details & Documents
Synonyms: Nonadecylic acid
- CAS Number:646-30-0
- Formula:C₁₉H₃₈O₂
- MDL Number:MFCD00002754
- Molecular Weight:298.51 g/mol
- Boiling Pt:297 °C (100 mmHg)
- Melting Pt:68…70 °C
- Density:0,8468 g/cm³ (70 °C)
- Storage Temperature:Ambient
- EINECS:211-472-4
Documents
ACRO20719
SDS
Specifications
Specification Test Results
- Appearance:White Glistening powder or flakes
- Infrared spectrum:Conforms
- GC:≥98.0 %
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



