Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Formamide 99.5% for analysis

Formamide 99.5% for analysis

Glass bottle, 100 mL
Catalog # ACRO205821000
Health
Formamide 99.5% for analysis
Catalog # ACRO205821000
CAS Number:  75-12-7
Formamide 99.5% for analysis
Formamide 99.5% for analysis

Glass bottle, 100 mL

CAS Number:  75-12-7
Price
27.30 SFr.
Log in for availability
1 * 100 mL
Product Details & Documents
Synonyms:  Carbamaldehyde
  • CAS Number:75-12-7
  • Formula:HCONH₂
  • MDL Number:MFCD00007941
  • Molecular Weight:45.04 g/mol
  • Merck Index:13,04261
  • Boiling Pt:210 °C (760 mmHg)
  • Melting Pt:2…3 °C
  • Density:1.133 g/cm³
  • Flash Pt:175 °C
  • Storage Temperature:Ambient
  • EINECS:200-842-0

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.05 % (Coulometric)
  • Sulfated ash:
    ≤0.005 %
  • GC:
    ≥99.5 %
  • Color scale:
    ≤10 APHA
  • Copper (Cu):
    ≤1 ppm
  • Iron (Fe):
    ≤1 ppm
  • Zinc (Zn):
    ≤1 ppm
  • Heavy metals (as Pb):
    ≤1 ppm

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.