Formamide 99.5% for analysis
Glass bottle, 2,5 L
Supplier: Thermo Fisher Scientific
Formamide 99.5% for analysis
Catalog # ACRO205820025
Supplier: Thermo Fisher Scientific
CAS Number: 75-12-7
Jump to:
Product Details & Documents
Synonyms: Carbamaldehyde
- CAS Number:75-12-7
- Formula:HCONH₂
- MDL Number:MFCD00007941
- Molecular Weight:45.04 g/mol
- Merck Index:13,04261
- Boiling Pt:210 °C (760 mmHg)
- Melting Pt:2…3 °C
- Density:1.133 g/cm³
- Flash Pt:175 °C
- Storage Temperature:Ambient
- EINECS:200-842-0
Specifications
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Water:≤0.05 % (Coulometric)
- Sulfated ash:≤0.005 %
- GC:≥99.5 %
- Color scale:≤10 APHA
- Copper (Cu):≤1 ppm
- Iron (Fe):≤1 ppm
- Zinc (Zn):≤1 ppm
- Heavy metals (as Pb):≤1 ppm
Specifications
- Pack type:Glass bottle
- Pk:2,5 L



