Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Bulk packaged serological pipettes, Falcon®

Bulk packaged serological pipettes, Falcon®

5 ml, 0,10 ml, +, 25 per bag
Catalog # 734-0344
Supplier:   Corning
Bulk packaged serological pipettes, Falcon®
Catalog # 734-0344
Supplier:  Corning
Bulk packaged serological pipettes, Falcon®
Bulk packaged serological pipettes, Falcon®

5 ml, 0,10 ml, +, 25 per bag

Price
SFr. 290.00
Log in for availability
1 * 500 ST
Product Details & Documents

The pipettes graduation alignment system is a fast and simple way to bring graduations to your line of sight. Polystyrene, disposable pipettes for tissue culture, bacteriological, and research applications.

  • Sterile
  • RNase-/DNase-free
  • Noncytotoxic, nonpyrogenicity tested to less than 0,1 EU/ml
  • Color-coded package and markings for ease of identification

Simply pop open or peel back the top of the pipette package and line up the pipettor before completely removing the pipette from its wrapping. When you remove the pipette from its package, the graduations should face you every time.

Specifications
  • Packed:
    25 per bag
  • Sterile:
    +
  • Capacity:
    5 ml
  • Seq:
    04
  • Division:
    0,10 ml