Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Cell strainers, Falcon®

Cell strainers, Falcon®

Cell strainers, 70 µm pore size, white frame
Catalog # 734-0003
Supplier:   Corning
Cell strainers, Falcon®
Catalog # 734-0003
Supplier:  Corning
Cell strainers, Falcon®
Cell strainers, Falcon®

Cell strainers, 70 µm pore size, white frame

Price
SFr. 161.00
Log in for availability
1 * 50 ST
Product Details & Documents

An easy, ready-to-use way to consistently obtain a more uniform single-cell suspension. Made of a strong nylon mesh and available with 40, 70 or 100 µm pores that are evenly spaced for optimal performance in a variety of applications. A faster and easier alternative to gauze filtration in procedures involving dissociation of cells from either clumps or primary tissue.

  • Moulded, colour-coded PP frame with tab enables easy handling
  • Fits perfectly into a 50 ml Falcon® conical tube or other similarly sized tube
  • Sterilised by gamma irradiation and conveniently available in individual, peel-open packaging
  • Extended lip on strainer enables aseptic handling with forceps

Ordering information: Packaged individually in bags.

Specifications
  • Description:
    Cell strainers, 70 µm pore size, white frame