Stearic acid 97%
Plastic bottle, 2,5 kg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Octadecanoic acid, Octadecylic acid
- CAS Number:57-11-4
- Formula:C₁₈H₃₆O₂
- MDL Number:MFCD00002752
- Molecular Weight:284.48 g/mol
- Merck Index:13,08882
- Boiling Pt:361 °C (760 mmHg)
- Melting Pt:67…69 °C
- Density:0.84 g/cm³
- Flash Pt:196 °C
- Storage Temperature:Ambient
- EINECS:200-313-4
Documents
ACRO17449
SDS
Specifications
Specification Test Results
- Appearance:White Granular powder or granules or flakes
- Infrared spectrum:Conforms
- Melting point:67 to 71 °C
- GC:≥96.0 %
Specifications
- Pack type:Plastic bottle
- Pk:2,5 kg



