Quercetin hydrate 95%
Glass bottle, 100 g
Supplier: Thermo Fisher Scientific
Quercetin hydrate 95%
Catalog # ACRO174071000
Supplier: Thermo Fisher Scientific
CAS Number: 849061-97-8
Jump to:
Product Details & Documents
Appearance: Yellow Crystalline powder
Synonyms: 3,3',4',5,7-Pentahydroxyflavone hydrate, 2-(3,4-Dihydroxyphenyl)-3,5,7-trihydroxy-4H-chromen-4-one xhydrate
- CAS Number:849061-97-8
- Formula:C₁₅H₁₀O₇·xH₂O
- MDL Number:MFCD00006828
- Molecular Weight:302.24 g/mol (anhy)
- Melting Pt:>300 °C
- Storage Temperature:Ambient
- UN:2811
- ADR:6.1,III
Documents
Specifications
Specification Test Results
- Appearance:Yellow Crystalline powder
- Infrared spectrum:Conforms
- Water:≤12 % (K.F.)
- HPLC:≥94 %
Specifications
- Pack type:Glass bottle
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



