5α-Cholestane 98+%
Glass bottle for solids, 1 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:481-21-0
- Formula:C₂₇H₄₈
- MDL Number:MFCD00066412
- Molecular Weight:372.68 g/mol
- Merck Index:12,02254
- Boiling Pt:250 °C (1 mmHg)
- Melting Pt:78…81 °C
- Density:0,9133 g/cm³ (80 °C)
- :Density: 0.9133 g/cm³ (80 °C)
- Storage Temperature:Ambient
- EINECS:207-562-8
Documents
ACRO16560
SDS
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:78 to 81 °C
- GC:≥98.0 %
- Specific optical rotation:+23° to +30° (20 °C, 589 nm) (c=2, CHCl3)
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



