Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

D-(-)-Fructose 99%

Plastic bottle, 1 kg
Catalog # ACRO161350010
D-(-)-Fructose 99%
Catalog # ACRO161350010
CAS Number:  57-48-7
D-(-)-Fructose 99%
D-(-)-Fructose 99%

Plastic bottle, 1 kg

CAS Number:  57-48-7
Price
SFr. 102.00
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  D(-)-Levulose, (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one, β-D-(-)-Fructose, Fructose
  • CAS Number:57-48-7
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00148910
  • Molecular Weight:180.16 g/mol
  • Merck Index:13,04295
  • Boiling Pt:552 °C (1013 hPa)
  • Melting Pt:103…105 °C (dec.)
  • Density:1,59 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-333-3
Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Assay:
    ≥98.5 %
  • Loss on drying:
    ≤0.5 % (70°C, 4 hrs) (Vacuum)
  • Sulfated ash:
    ≤0.1 %
  • Additional info:
    5-Hydroxymethylfurfural and rel. compounds: Passes
  • Appearance of solution:
    Passes test
  • Specific optical rotation:
    -91.0° to -93.5° (20°C, 589 nm) (c=10, H2O/NH3) (After 30 min, On dry substance)
  • Chloride (Cl):
    ≤50 ppm
  • Sulfate (SO4):
    ≤50 ppm
  • Heavy metals (as Pb):
    ≤10 ppm
  • Foreign sugars:
    Passes test

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 kg

Specifications