n-Hexane 99+% for analysis
Glass bottle, 2,5 L
Supplier: Thermo Fisher Scientific
n-Hexane 99+% for analysis
Catalog # ACRO160780025
Supplier: Thermo Fisher Scientific
CAS Number: 110-54-3
Jump to:
Product Details & Documents
Synonyms: Hexyl hydride, Hexanen, Hexane
- CAS Number:110-54-3
- Formula:H₃C(CH₂)₄CH₃
- MDL Number:MFCD00009520
- Molecular Weight:86.18 g/mol
- Merck Index:13,04712
- Boiling Pt:69 °C (760 mmHg)
- Melting Pt:–95 °C
- Density:0.659 g/cm³
- Flash Pt:–22 °C
- Storage Temperature:Ambient
- EINECS:203-777-6
- UN:1208
- ADR:3,II
Documents
ACRO16078
SDS
Thermo Scientific Stockroom Essentials
Product Literature
Specifications
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.02 % (Coulometric)
- GC:≥99.0 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.002 %
- Calcium (Ca):≤0.5 ppm
- Copper (Cu):≤0.1 ppm
- Iron (Fe):≤0.1 ppm
- Lead (Pb):≤0.1 ppm
- Magnesium (Mg):≤0.1 ppm
- Zinc (Zn):≤0.1 ppm
- Sulfur compounds (as S):≤0.005 %
- Free acid (CH3COOH):≤0.002 %
- Thiophene:Passes test
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



