96-Well clear polystyrene microplates, Corning®
Standard plate, V bottom, non treated, without lid*, 96, Clear, 75 - 200 µl, 320 µl, -, 25/bag
Supplier: Corning
96-Well clear polystyrene microplates, Corning®
Catalog # 734-1659
Supplier: Corning
Jump to:
Product Details & Documents
Cell culture microplates are sterile and nonpyrogenic. Lids available where indicated.
Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays. Cat. Nos. 3585 and 3595 have special low evaporation lids. Cat. No. 3841 is aseptically manufactured.
Specifications
- No. of wells:96
- Packed:25/bag
- Recommended working volume:75 - 200 µl
- Sterile:-
- Colour:Clear
- Description:Standard plate, V bottom, non treated, without lid*
- Seq:37
- Well volume:320 µl
- Cat. No.:734-1659
- Environmental certification:RECs, ISO 14001 - Kennebunk
- Environmentally Preferable:Low Manufacturing Impact



