Clear inserts, polyester (PET) membrane, Transwell®
6-well insert plates, 4,67 cm², Polyester, 24 mm, 3,0 µm
Supplier: Corning
Clear inserts, polyester (PET) membrane, Transwell®
Catalog # 734-1578
Supplier: Corning
Jump to:
Product Details & Documents
Permeable supports, also known as cell culture inserts, are an essential tool for the study of both anchorage-dependent and independent cell lines.
- w Produce a cell culture environment that closely resembles an in vivo state
- Co-culture cells with or without cell-to-cell contact
- Design a diversity of experiments using various pore sizes, membrane types, and coatings 10 µm transparent membrane
- Treated for optimal cell attachment
- Excellent visibility under phase contrast microscopy
- Supplied in multiple well plates
- Sterile
Allow polarised cells to carry out metabolic activities in a more natural manner because the cells feed both apically and basolaterally.
Dokumente
Specifications
- Membrane:Polyester
- Pore size:3,0 µm
- Description:6-well insert plates
- Seq:07
- Culture area:4,67 cm²
- Membrane Ø:24 mm
- Cat. No.:734-1578
- Environmental certification:RECs, ISO 14001 - Kennebunk
- Environmentally Preferable:Low Manufacturing Impact
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen..



