96-Well clear polystyrene microplates, Corning®
Standard plate, Easy Wash™, High Bind surface, without lid, for EIA/RIA, 96, Clear, 75 - 200 µl, 360 µl, -, 25/bag
Supplier: Corning
96-Well clear polystyrene microplates, Corning®
Catalog # 734-1786
Supplier: Corning
Jump to:
Product Details & Documents
Cell culture microplates are sterile and nonpyrogenic. Lids available where indicated.
Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays. Cat. Nos. 3585 and 3595 have special low evaporation lids. Cat. No. 3841 is aseptically manufactured.
Specifications
- No. of wells:96
- Packed:25/bag
- Recommended working volume:75 - 200 µl
- Sterile:-
- Colour:Clear
- Description:Standard plate, Easy Wash™, High Bind surface, without lid, for EIA/RIA
- Seq:08
- Well volume:360 µl
- Cat. No.:734-1786
- Environmental certification:RECs, ISO 14001 - Kennebunk
- Environmentally Preferable:Low Manufacturing Impact



