Order Entry
Switzerland
Orders LinkContactUsLinkComponent
4-Nitrophenol 99%

4-Nitrophenol 99%

Glass bottle for solids, 5 g
Catalog # ACRO157050050
Toxic
Health
4-Nitrophenol 99%
Catalog # ACRO157050050
CAS Number:  100-02-7
4-Nitrophenol 99%
4-Nitrophenol 99%

Glass bottle for solids, 5 g

CAS Number:  100-02-7
Price
SFr. 38.50
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  1-Hydroxy-4-nitrobenzene, p-Nitrophenol
  • CAS Number:100-02-7
  • Formula:C₆H₅NO₃
  • MDL Number:MFCD00007331
  • Molecular Weight:139.11 g/mol
  • Merck Index:13,06654
  • Boiling Pt:279 °C (760 mmHg)
  • Melting Pt:>112 °C
  • Density:1,48 g/cm³ (20 °C)
  • Flash Pt:169 °C
  • Storage Temperature:Ambient
  • EINECS:202-811-7
  • UN:1663
  • ADR:6.1,III

Documents

Specifications

Specification Test Results

  • Appearance:
    Yellow to brown Crystalline powder or crystals and/or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    112 to 115 °C
  • GC:
    ≥98.5 %
  • Titration with NaOH:
    ≥96.0 to ≤104.0 %

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.