Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Thermometer probes

Thermometer probes

Food penetration probe with thermistor, blue cap with cable (1 m), PX25L, −40...+150 °C, 3,3×100 mm
Catalog # 620-0365
Supplier:   COMARK
Thermometer probes
Catalog # 620-0365
Supplier:  COMARK
Thermometer probes

Food penetration probe with thermistor, blue cap with cable (1 m), PX25L, −40...+150 °C, 3,3×100 mm

Price
SFr. 115.00
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Range:
    −40...+150 °C
  • Description:
    Food penetration probe with thermistor, blue cap with cable (1 m), PX25L
  • Seq:
    05
  • Ø×L:
    3,3×100 mm