Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Thermometer probes

Thermometer probes

Food penetration probe with thermistor, green cap with cable (1 m), PX24L, −40...+150 °C, 3,3×100 mm
Catalog # 620-0364
Supplier:   COMARK
Thermometer probes
Catalog # 620-0364
Supplier:  COMARK
Thermometer probes

Food penetration probe with thermistor, green cap with cable (1 m), PX24L, −40...+150 °C, 3,3×100 mm

Price
SFr. 81.70
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Range:
    −40...+150 °C
  • Description:
    Food penetration probe with thermistor, green cap with cable (1 m), PX24L
  • Seq:
    04
  • Ø×L:
    3,3×100 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.