Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Caprinaldehyde 95%

Caprinaldehyde 95%

Glass bottle, 100 mL
Catalog # ACRO154971000
Irritant
Caprinaldehyde 95%
Catalog # ACRO154971000
CAS Number:  112-31-2
Caprinaldehyde 95%
Caprinaldehyde 95%

Glass bottle, 100 mL

CAS Number:  112-31-2
Price
SFr. 34.70
Log in for availability
1 * 100 mL
Product Details & Documents
Synonyms:  Decylaldehyde, Decanal, Decyl aldehyde
  • CAS Number:112-31-2
  • Formula:C₁₀H₂₀O
  • MDL Number:MFCD00007031
  • Boiling Pt:207…209 °C (760 mmHg)
  • Density:0.83 g/cm³
  • Storage Temperature:Ambient
  • EINECS:203-957-4

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.