4-Nitro-o-anisidine 99%
Glass bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
4-Nitro-o-anisidine 99%
Catalog # ACRO150640050
Supplier: Thermo Fisher Scientific
CAS Number: 97-52-9
Jump to:
Product Details & Documents
Synonyms: 2-Methoxy-4-nitroaniline, 2-Amino-5-nitroanisole
- CAS Number:97-52-9
- Formula:C₇H₈N₂O₃
- MDL Number:MFCD00007363
- Molecular Weight:168.15 g/mol
- Boiling Pt:364 °C (1013 hPa)
- Melting Pt:139…142 °C
- Density:1,2112 g/cm³ (156 °C)
- Storage Temperature:Ambient
- EINECS:202-588-6
Documents
ACRO15064
SDS
Specifications
Specification Test Results
- Appearance:Yellow to orange Crystalline powder
- Infrared spectrum:Conforms
- Melting point:139 to 142 °C
- GC:≥98.5 %
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



