Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Propionic acid 99%, pure

Propionic acid 99%, pure

Glass bottle, 2,5 L
Catalog # ACRO149300025
Corrosive
Flammable
Irritant
Propionic acid 99%, pure
Catalog # ACRO149300025
CAS Number:  79-09-4
Propionic acid 99%, pure
Propionic acid 99%, pure

Glass bottle, 2,5 L

CAS Number:  79-09-4
Price
SFr. 87.70
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  Propanoic acid
  • CAS Number:79-09-4
  • Formula:C₃H₆O₂
  • MDL Number:MFCD00002756
  • Molecular Weight:74.08 g/mol
  • Merck Index:13,07917
  • Boiling Pt:141 °C (760 mmHg)
  • Melting Pt:–22 °C
  • Density:0.99 g/cm³
  • Flash Pt:51 °C
  • Storage Temperature:Ambient
  • EINECS:201-176-3
  • UN:3463
  • ADR:8,II
  • REACH:01-2119486971-24

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3850 to 1.3870 (20°C, 589 nm)
  • Water:
    ≤0.2 %
  • GC:
    ≥98.5 %
  • Color scale:
    ≤10 APHA

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.