ContactUsLinkComponent
L5-well PTFE Comb for SE 250/SE 260 Mini-Vertical Gel Units 13 x 1.00mm (W x T) 1 * 1 items
Catalog # SE211A-5-1.0
Supplier:  Cytiva
L5-well PTFE Comb for SE 250/SE 260 Mini-Vertical Gel Units 13 x 1.00mm (W x T) 1 * 1 items
Catalog # SE211A-5-1.0
Supplier:  Cytiva