Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Tridecanoic acid 97%

Tridecanoic acid 97%

Glass bottle for solids, 25 g
Catalog # ACRO139520250
Irritant
Tridecanoic acid 97%
Catalog # ACRO139520250
CAS Number:  638-53-9
Tridecanoic acid 97%
Tridecanoic acid 97%

Glass bottle for solids, 25 g

CAS Number:  638-53-9
Price
SFr. 185.00
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  n-Tridecanoic acid, Tridecylic acid
  • CAS Number:638-53-9
  • Formula:C₁₃H₂₆O₂
  • MDL Number:MFCD00002741
  • Molecular Weight:214.35 g/mol
  • Boiling Pt:236 °C (100 mmHg)
  • Melting Pt:40…45 °C
  • Density:1,025 g/cm³ (20 °C)
  • Flash Pt:113 °C
  • Storage Temperature:Ambient
  • EINECS:211-341-1

Documents

Specifications

Specification Test Results

  • Appearance:
    White to light yellow Crystalline powder or flakes and/or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    40 to 45 °C
  • Titration with NaOH:
    ≥96.0 %

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.