Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Tridecanoic acid 97%

Tridecanoic acid 97%

Glass bottle, 10 g
Catalog # ACRO139520100
Irritant
Tridecanoic acid 97%
Catalog # ACRO139520100
CAS Number:  638-53-9
Tridecanoic acid 97%
Tridecanoic acid 97%

Glass bottle, 10 g

CAS Number:  638-53-9
Price
91.10 SFr.
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  n-Tridecanoic acid, Tridecylic acid
  • CAS Number:638-53-9
  • Formula:C₁₃H₂₆O₂
  • MDL Number:MFCD00002741
  • Molecular Weight:214.35 g/mol
  • Boiling Pt:236 °C (100 mmHg)
  • Melting Pt:40…45 °C
  • Density:1,025 g/cm³ (20 °C)
  • Flash Pt:113 °C
  • Storage Temperature:Ambient
  • EINECS:211-341-1

Documents

Specifications

Specification Test Results

  • Appearance:
    White to light yellow Crystalline powder or flakes and/or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    40 to 45 °C
  • Titration with NaOH:
    ≥96.0 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 g

Specifications