Pentadecanoic acid 99%
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
Pentadecanoic acid 99%
Catalog # ACRO129830250
Supplier: Thermo Fisher Scientific
CAS Number: 1002-84-2
Jump to:
Product Details & Documents
Synonyms: Pentadecylic acidn-Pentadecanoic acid
- CAS Number:1002-84-2
- Formula:C₁₅H₃₀O₂
- MDL Number:MFCD00002745
- Molecular Weight:242.4 g/mol
- Boiling Pt:339 °C (1013 hPa)
- Melting Pt:51…53 °C
- Density:0,8496 g/cm³ (70 °C)
- Flash Pt:>110 °C
- Storage Temperature:Ambient
- EINECS:213-693-1
Documents
Specifications
Specification Test Results
- Appearance:White to off-white Powder or crystals or flakes and/or chunks
- Infrared spectrum:Conforms
- GC:≥98.5 %
- Titration with NaOH:≥98.5 %
- Specific optical rotation:-63° to -67° (20°C, 589 nm) (c=4, methanol)
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



