ContactUsLinkComponent
MMP-13, Human, Biotrak Assay, ELISA - 96 wells (Specific for MMP-13. Both pro- and active MMP-13 is detected. Range: 0.094-3ng/ml. S ensitivity: 0.032ng/ml) 1 * 1 items
Catalog # RPN2621
Supplier:  Cytiva
MMP-13, Human, Biotrak Assay, ELISA - 96 wells (Specific for MMP-13. Both pro- and active MMP-13 is detected. Range: 0.094-3ng/ml. S ensitivity: 0.032ng/ml) 1 * 1 items
Catalog # RPN2621
Supplier:  Cytiva
Frequently Bought Together