4-Methylmorpholine 99%
Glass bottle for solids, 500 g
Supplier: Thermo Fisher Scientific
4-Methylmorpholine 99%
Catalog # ACRO127155000
Supplier: Thermo Fisher Scientific
CAS Number: 109-02-4
Jump to:
Product Details & Documents
Synonyms: NMM, N-Methylmorpholine
- CAS Number:109-02-4
- Formula:C₅H₁₁NO
- MDL Number:MFCD00006175
- Molecular Weight:101.15 g/mol
- Boiling Pt:115…116 °C (750 mmHg)
- Melting Pt:–65 °C
- Density:0.92 g/cm³
- Flash Pt:14 °C
- Storage Temperature:Ambient
- EINECS:203-640-0
- UN:2535
- ADR:3,II
Specifications
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4339 to 1.4359 (20°C, 589 nm)
- Water:≤0.5 % (K.F.)
- GC:≥98.5 %
- Color scale:≤50 APHA
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g



