Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Caproic acid 99%

Caproic acid 99%

Glass bottle for solids, 1 kg
Catalog # ACRO120700010
Corrosive
Caproic acid 99%
Catalog # ACRO120700010
CAS Number:  142-62-1
Caproic acid 99%
Caproic acid 99%

Glass bottle for solids, 1 kg

CAS Number:  142-62-1
Price
91.50 SFr.
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Hexanoic acid
  • CAS Number:142-62-1
  • Formula:C₆H₁₂O₂
  • MDL Number:MFCD00004421
  • Molecular Weight:116.16 g/mol
  • Merck Index:13,01765
  • Boiling Pt:202…203 °C (760 mmHg)
  • Melting Pt:–3 °C
  • Density:0.92 g/cm³
  • Flash Pt:104 °C
  • Storage Temperature:Ambient
  • EINECS:205-550-7
  • UN:2829
  • ADR:8,III

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4150 to 1.4180 (20°C, 589 nm)
  • GC:
    ≥98.5 %
  • Acid value:
    473 to 493 mg KOH/g
  • test 11:
    Result 11
  • test 12:
    Result 12
  • test 13:
    Result 13
  • test 14:
    Result 14
  • test 15:
    Result 15
  • test 16:
    Result 16
  • test 17:
    Result 17
  • test 18:
    Result 18
  • test 19:
    Result 19
  • test 20:
    Result 20
  • test 21:
    Result 21
  • test 22:
    Result 22
  • test 23:
    Result 23
  • test 24:
    Result 24
  • test 25:
    Result 25
  • test 26:
    Result 26
  • test 27:
    Result 27
  • test 28:
    Result 28
  • test 29:
    Result 29
  • test 30:
    Result 30
  • test 31:
    Result 31
  • test 32:
    Result 32
  • test 33:
    Result 33
  • test 34:
    Result 34
  • test 35:
    Result 35
  • test 36:
    Result 36
  • test 37:
    Result 37
  • test 38:
    Result 38
  • test 39:
    Result 39
  • test 40:
    Result 40
  • test 41:
    Result 41
  • test 42:
    Result 42
  • test 43:
    Result 43
  • test 44:
    Result 44
  • test 45:
    Result 45
  • test 46:
    Result 46
  • test 47:
    Result 47
  • test 48:
    Result 48
  • test 49:
    Result 49
  • test 50:
    Result 50
  • test 51:
    Result 51
  • test 52:
    Result 52
  • test 53:
    Result 53
  • test 54:
    Result 54
  • test 55:
    Result 55
  • test 56:
    Result 56
  • test 57:
    Result 57

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.