ContactUsLinkComponent
TNFalpha, Mouse, Biotrak Assay, ELISA - 96 wells (Range: 43-2000pg/ml. Sensitivity: 10pg/ml. Sample volume: 50ul. 1.5L wash buffer supplied. Suitable for use with cell-culture supernatant and serum samples) 1 * 96 Assays
Catalog # RPN2718
Supplier:  Cytiva
TNFalpha, Mouse, Biotrak Assay, ELISA - 96 wells (Range: 43-2000pg/ml. Sensitivity: 10pg/ml. Sample volume: 50ul. 1.5L wash buffer supplied. Suitable for use with cell-culture supernatant and serum samples) 1 * 96 Assays
Catalog # RPN2718
Supplier:  Cytiva
Frequently Bought Together