Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Cyclohexanone 99.8%, extra pure

Cyclohexanone 99.8%, extra pure

Glass bottle, 2,5 L
Catalog # ACRO111190025
Irritant
Corrosive
Flammable
   Safer Human and Environmental Health
Cyclohexanone 99.8%, extra pure
Catalog # ACRO111190025
   Safer Human and Environmental Health
CAS Number:  108-94-1
Cyclohexanone 99.8%, extra pure
Cyclohexanone 99.8%, extra pure

Glass bottle, 2,5 L

CAS Number:  108-94-1
Price
SFr. 185.00
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  Pimelic Ketone
  • CAS Number:108-94-1
  • Formula:C₆H₁₀(=O)
  • MDL Number:MFCD00001625
  • Molecular Weight:98.14 g/mol
  • Merck Index:13,02755
  • Boiling Pt:155 °C (760 mmHg)
  • Melting Pt:–47 °C
  • Density:0.947 g/cm³
  • Flash Pt:43 °C
  • Storage Temperature:Ambient
  • EINECS:203-631-1
  • UN:1915
  • ADR:3,III
  • REACH:01-2119453616-35

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4450 to 1.4520 (20°C, 589 nm)
  • Water:
    ≤0.2 % (K.F.)
  • GC:
    ≥99.75 %
  • Color scale:
    ≤20 APHA
  • Iron (Fe):
    ≤0.001 %
  • Magnesium (Mg):
    ≤0.005 %
  • Manganese (Mn):
    ≤0.002 %
  • Potassium (K):
    ≤0.01 %
  • Sodium (Na):
    ≤0.05 %
  • Insoluble matter:
    ≤0.005 %
  • Nitrogen compounds (as N):
    ≤0.002 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Cat. No.:
    ACRO111190025
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default
  • Environmentally Preferable:
    Safer Human and Environmental Health

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.