Cyclohexanone 99.8%, extra pure
Glass bottle, 1 L
Supplier: Thermo Fisher Scientific
Cyclohexanone 99.8%, extra pure
Catalog # ACRO111190010
Supplier: Thermo Fisher Scientific
CAS Number: 108-94-1
Jump to:
Product Details & Documents
Synonyms: Pimelic Ketone
- CAS Number:108-94-1
- Formula:C₆H₁₀(=O)
- MDL Number:MFCD00001625
- Molecular Weight:98.14 g/mol
- Merck Index:13,02755
- Boiling Pt:155 °C (760 mmHg)
- Melting Pt:–47 °C
- Density:0.947 g/cm³
- Flash Pt:43 °C
- Storage Temperature:Ambient
- EINECS:203-631-1
- UN:1915
- ADR:3,III
- REACH:01-2119453616-35
Documents
Specifications
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4450 to 1.4520 (20°C, 589 nm)
- Water:≤0.2 % (K.F.)
- GC:≥99.75 %
- Color scale:≤20 APHA
- Iron (Fe):≤0.001 %
- Magnesium (Mg):≤0.005 %
- Manganese (Mn):≤0.002 %
- Potassium (K):≤0.01 %
- Sodium (Na):≤0.05 %
- Insoluble matter:≤0.005 %
- Nitrogen compounds (as N):≤0.002 %
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Cat. No.:ACRO111190010
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
- Environmentally Preferable:Safer Human and Environmental Health
Specifications
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



