ContactUsLinkComponent
Thermometer, cylinder, in tinned wire frame, mercury filled, blue reflecting, measuring range:0:150/1C, total immersion, capillary lens front, dimension:300x30mm. Until 10th October 2017 it is still allowed to use mercury thermometers if they are e 1 * 1 items
Catalog # AMART108162
Supplier:  Amarell
Thermometer, cylinder, in tinned wire frame, mercury filled, blue reflecting, measuring range:0:150/1C, total immersion, capillary lens front, dimension:300x30mm. Until 10th October 2017 it is still allowed to use mercury thermometers if they are e 1 * 1 items
Catalog # AMART108162
Supplier:  Amarell
Frequently Bought Together