ContactUsLinkComponent
Thermometer, precision, mercury filled, blue reflecting, measuring range:-10:50/0.1°C, with mercury protection, PTFE coated, dimens i on: 420x8-9mm, enclosed scale, capillary lens front, total immersion, round richter-top finish, environmentally, su 1 * 1 items
Katalognr. 620-1216
Supplier:  Amarell
undefined
Thermometer, precision, mercury filled, blue reflecting, measuring range:-10:50/0.1°C, with mercury protection, PTFE coated, dimens i on: 420x8-9mm, enclosed scale, capillary lens front, total immersion, round richter-top finish, environmentally, su 1 * 1 items
Katalognr. 620-1216
Supplier:  Amarell
Frequently Bought Together