ContactUsLinkComponent
Thermometer, Anschutz milk glass type set, measuring range:-10:360/0.1C, enclosed scale, 7 thermometers in stuffed case, governmen t verified, with verification vertificate or government calibrated, with calibration certificate. Until 10th October 1 * 1 items
Catalog # AMARN60155
Supplier:  Amarell
Thermometer, Anschutz milk glass type set, measuring range:-10:360/0.1C, enclosed scale, 7 thermometers in stuffed case, governmen t verified, with verification vertificate or government calibrated, with calibration certificate. Until 10th October 1 * 1 items
Catalog # AMARN60155
Supplier:  Amarell
Frequently Bought Together