ContactUsLinkComponent
Thermometer, precision, mercury filled, blue reflecting, DIN certified, measuring range:-10:50/0.1C, enclosed scale, capllary lens front, colourless, Richter-top finish with button, dimension: 420x8.5-8mm, suitable for government verification. Unti 1 * 1 items
Catalog # 620-1316
Supplier:  Amarell
Thermometer, precision, mercury filled, blue reflecting, DIN certified, measuring range:-10:50/0.1C, enclosed scale, capllary lens front, colourless, Richter-top finish with button, dimension: 420x8.5-8mm, suitable for government verification. Unti 1 * 1 items
Catalog # 620-1316
Supplier:  Amarell
Frequently Bought Together