Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Twaddle hydrometers

Twaddle hydrometers

Twaddle hydrometer, 102 – 138 °TW, 0,5 °TW, 280 mm
Catalog # 614-1625
Supplier:   Alla France
Twaddle hydrometers
Catalog # 614-1625
Supplier:  Alla France
Twaddle hydrometers
Twaddle hydrometers

Twaddle hydrometer, 102 – 138 °TW, 0,5 °TW, 280 mm

Price
SFr. 54.50
Log in for availability
1 * 1 ST
Product Details & Documents

Hydrometers for sugar with scales.

Calibration temperature of 15.6 °C.

Specifications
  • Description:
    Twaddle hydrometer
  • Length:
    280 mm
  • Division:
    0,5 °TW
  • Range:
    102 – 138 °TW
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.