Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Twaddle hydrometers

Twaddle hydrometers

Twaddle hydrometer, 24 – 48 °TW, 0,5 °TW, 280 mm
Catalog # 614-1622
Supplier:   Alla France
Twaddle hydrometers
Catalog # 614-1622
Supplier:  Alla France
Twaddle hydrometers
Twaddle hydrometers

Twaddle hydrometer, 24 – 48 °TW, 0,5 °TW, 280 mm

Price
SFr. 32.70
Log in for availability
1 * 1 ST
Product Details & Documents

Hydrometers for sugar with scales.

Calibration temperature of 15.6 °C.

Specifications
  • Description:
    Twaddle hydrometer
  • Length:
    280 mm
  • Division:
    0,5 °TW
  • Range:
    24 – 48 °TW